missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB1 Antibody (OTI4E12), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72573PE
This item is not returnable.
View return policy
Description
MAGEB1 Monoclonal antibody specifically detects MAGEB1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MAGEB1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4112 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4E12 | |
| Western Blot, Immunohistochemistry | |
| Cancer/testis antigen 3.1, CT3.1cancer/testis antigen family 3, member 1, DAM10MAGE-B1 antigen, DSS/AHC critical interval MAGE superfamily 10, DSS-AHC critical interval MAGE superfamily 10, MAGEL1MAGE-XP antigen, MAGE-Xp, MAGEXP, melanoma antigen family B, 1, melanoma-associated antigen B1, MGC9322 | |
| Full length human recombinant protein of human MAGEB1 (NP_796379) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction