missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551744
This item is not returnable.
View return policy
Description
Immunogen sequence: LKVAHATAAE KEECPSSSPV LGDTPTSSPA AGIPQKPQGA PPTTTAAAAV SCTESDEGAK CQGEENASFS QATTSTESSV KDPVAWEAGM LMHFILRKYK MREPIMKADM LKVVDEKYKD HFTEILNGAS R Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 38%.
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region, and expressed in testis and in a significant fraction of tumors of various histological types. This gene and other MAGEB members are clustered on chromosome Xp22-p21. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene, however, the full length nature of some variants has not been defined.
Specifications
| MAGEB1 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB1 | |
| cancer/testis antigen 3.1; cancer/testis antigen family 3, member 1; CT3.1; dam1; DAM10; DSS/AHC critical interval MAGE superfamily 10; DSS-AHC critical interval MAGE superfamily 10; LOC491803; MAGE family member B1; MAGEB1; Mage-b1; MAGE-B1 antigen; Magel1; Mage-rs1; MAGEXP; MAGE-Xp; MAGE-XP antigen; melanoma antigen family B, 1; melanoma antigen family B1; melanoma antigen, family B, 1; melanoma-associated antigen B1; MGC9322; RGD1565746; Smage1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43366 | |
| MAGEB1 | |
| Recombinant protein corresponding to Human MAGEB1. Recombinant protein control fragment (Product #RP-96775). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction