missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEB17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552001
This item is not returnable.
View return policy
Description
Immunogen sequence: LYLKEMDPSR QSYVLVGKLD FPNQGSLSDG GGFPLSGLLM VLLSTIFMHG NRATEEEMWE CLNALGMYKG RKHFIYGEPQ ELVTKDLVRE GYLEYQQVPS SDPPRYEFLW GPRARAETSK MKV Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 64%.
MAGEB17 is a protein coding gene.
Specifications
| MAGEB17 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB17 | |
| MAGE family member B17; MAGEB17; melanoma antigen family B, 17 (pseudogene); melanoma antigen family B17; melanoma-associated antigen B17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 645864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A8MXT2 | |
| MAGEB17 | |
| Recombinant protein corresponding to Human MAGEB17. Recombinant protein control fragment (Product #RP-92256). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction