missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB18 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502493
This item is not returnable.
View return policy
Description
The melanoma-associated antigen (MAGE) family consists of a number of antigens recognized by cytotoxic T lymphocytes. The MAGE genes were initially isolated from different kinds of tumors and, based on their virtually exclusive tumor-specific expression in adult tissues, they have been used as targets for cancer immunotherapy. MAGE genes encode for tumor-rejection antigens that are expressed in tumors of different histologic types and in normal testis and placenta. MAGE-B18 (melanoma-associated antigen B18) is a 343 amino acid protein that contains one MAGE domain and may be involved in tumorigenesis. The gene encoding MAGE-B18 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.Specifications
| MAGEB18 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96M61 | |
| MAGEB18 | |
| Full length human recombit protein of human MAGEB18 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1F5 | |
| Unconjugated | |
| MAGEB18 | |
| MAGE family member B18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 286514 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction