missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB18 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502493S
This item is not returnable.
View return policy
Description
MAGEB18 Monoclonal antibody specifically detects MAGEB18 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MAGEB18 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50552 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7408 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1F5 | |
| Unconjugated | |
| VASP | |
| MAGBI; MAGE family member B18; MAGEB18; MAGE-B18 antigen; melanoma antigen family B, 18; melanoma antigen family B18; melanoma-associated antigen B18 | |
| Full length human recombinant protein of human VASP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction