missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550825
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MAGEB2 protein.
MAGEB2 is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
Specifications
| MAGEB2 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB2 | |
| Cancer/testis antigen 3.2; cancer/testis antigen family 3, member 2; CT3.2; DAM6; DSS/AHC critical interval MAGE superfamily 6; DSS-AHC critical interval MAGE superfamily 6; MAGE family member B2; MAGE XP-2 antigen; MAGEB2; MAGE-B2 antigen; MAGE-XP-2; melanoma antigen family B, 2; melanoma antigen family B2; melanoma-associated antigen B2; MGC26438 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4113 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O15479 | |
| MAGEB2 | |
| Synthetic peptide corresponding to residues near the N terminal of human melanoma antigen family B, 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction