missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58828
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MAGEB3 Polyclonal specifically detects MAGEB3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| MAGEB3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CT3.5, MAGE-B3 antigen, melanoma antigen family B, 3, melanoma-associated antigen B3, member 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4114 | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MAGEB3 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TRGQTQDHQGAQITATNKKKVSFSSPLILGATIQKKSAGRSRSALKKPQRA | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu