Learn More
MAGEB3 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808753
Encompass_Preferred
 View more versions of this product
Product Code. 17580334
  • € 540.87 / 100µL
Description
This gene is a MAGE-B subfamily member of the MAGE gene family. MAGE family member proteins direct the expression of tumor antigens recognized on a human melanoma by autologous cytolytic T lymphocytes. There are two known clusters of MAGE genes on chromosome X. The members of the MAGE-A subfamily are located in the Xq28 region, while the members of the MAGE-B subfamily are clustered in the Xp21 region.
Specifications
Specifications
MAGEB3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O15480
MAGEB3
Full length human recombit protein of human MAGEB3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1D3
Unconjugated
MAGEB3
CT3.5
Mouse
Affinity Chromatography
RUO
4114
-20° C, Avoid Freeze/Thaw Cycles