missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAGEB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15812814
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15812814 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15812814 Supplier Invitrogen™ Supplier No. PA567683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGEB3 Polyclonal Antibody for Western Blot

This gene is a MAGE-B subfamily member of the MAGE gene family. MAGE family member proteins direct the expression of tumor antigens recognized on a human melanoma by autologous cytolytic T lymphocytes. There are two known clusters of MAGE genes on chromosome X. The members of the MAGE-A subfamily are located in the Xq28 region, while the members of the MAGE-B subfamily are clustered in the Xp21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEB3
Gene Accession No. O15480
Gene Alias cancer/testis antigen family 3, member 5; CT3.5; LOW QUALITY PROTEIN: melanoma-associated antigen B3; MAGE family member B3; MAGEB3; Mage-b3; MAGE-B3 antigen; Mage-ps1; Mage-rs3; melanoma antigen family B, 3; melanoma antigen family B3; melanoma antigen, family B, 3; melanoma-associated antigen B3; RGD1562343; Smage3
Gene Symbols MAGEB3
Host Species Rabbit
Immunogen Synthesized peptide derived from human MAGEB3 (aa 1-28).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4114
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.