missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570209
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Dog: 77%; Guinea Pig: 77%; Horse: 77%; Human: 100%; Mouse: 77%; Rabbit: 79%; Rat: 77%.
This gene is a MAGE-B subfamily member of the MAGE gene family. MAGE family member proteins direct the expression of tumor antigens recognized on a human melanoma by autologous cytolytic T lymphocytes. There are two known clusters of MAGE genes on chromosome X. The members of the MAGE-A subfamily are located in the Xq28 region, while the members of the MAGE-B subfamily are clustered in the Xp21 region.
Specifications
| MAGEB3 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB3 | |
| cancer/testis antigen family 3, member 5; CT3.5; LOW QUALITY PROTEIN: melanoma-associated antigen B3; MAGE family member B3; MAGEB3; Mage-b3; MAGE-B3 antigen; Mage-ps1; Mage-rs3; melanoma antigen family B, 3; melanoma antigen family B3; melanoma antigen, family B, 3; melanoma-associated antigen B3; RGD1562343; Smage3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4114 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15480 | |
| MAGEB3 | |
| synthetic peptide directed towards the N terminal region of human MAGEB3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction