Learn More
MAGEB4 Mouse anti-Human, Clone: OTI3A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809181
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.Specifications
| MAGEB4 | |
| Monoclonal | |
| Unconjugated | |
| MAGEB4 | |
| CT3.6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4115 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15481 | |
| MAGEB4 | |
| Human recombit protein fragment corresponding to amino acids 1-193 of human MAGEB4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.