missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGED1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MAGED1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878683
|
Proteintech
11539-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868683
|
Proteintech
11539-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGE-D2, also known as Melanoma-associated antigen D2, belongs to the melanoma antigen encoding gene (MAGE) family with a MAGE domain. Rat and mouse orthologues have been isolated. Compared to other proteins (MAGE-A, MAGE- B and MAGE-C clusters) MAGE-D2 is expressed ubiquitously. This intracellular protein shows high expression in specific brain regions and in the interstitium of testes. Though the exact function of this protein is yet to be deduced, MAGE-D2 is considered as a hot spot for X-linked mental retardation. Research reveals that MAGE-D2 might have some role in breast and gastric cancer, as well as in hepatocellular carcinoma. MAGE-D2 is also known to interact physically with p53 and modify its activity, thus acting as a p53-dissociator.Specifications
| MAGED1 | |
| Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q9ES73, Q9QYH6, Q9Y5V3 | |
| 84469, 94275, 9500 | |
| MAGED1 Fusion Protein Ag2109 | |
| Primary | |
| -20°C | |
| MAGED1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DLXIN 1, MAGE D1 antigen, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, Melanoma associated antigen D1, NRAGE, PP2250 | |
| Maged1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title