missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102024
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAGEC2.
MAGE-C2/C3 (Melanoma-associated antigen C2, Hepatocellular carcinoma-associated antigen 587) is a 373 amino acid protein encoded by the human gene MAGEC2. Genes of the MAGE family direct the expression of tumor antigens that are recognized on human melanomas by autologous cytolytic T lymphocytes. Among tumor tissues, MAGE-C2/C3 is frequently expressed in seminomas, melanomas, and bladder transitional cell carcinomas. It is also expressed in a significant fraction of head and neck carcinomas, breast carcinomas, nonsmall-cell lung carcinomas, and sarcomas. In hepatocellular carcinoma, there is a inverse correlation between tumor differentiation and protein expression, i. e. the lower the differentiation, the higher percentage of expression. MAGE-C2/C3 is not expressed in normal tissues, except in germ cells in the seminiferous tubules and in Purkinje cells of the cerebellum. It is strongly expressed in spermatogonia and primary spermatocytes, however, at later stages of maturation, expression gradually decreases to become undetectable in mature spermatids.
Specifications
| MAGEC2 | |
| Polyclonal | |
| Unconjugated | |
| MAGEC2 | |
| Cancer/testis antigen 10; CT10; HCA587; hepatocellular cancer antigen 587; hepatocellular carcinoma-associated antigen 587; MAGE family member C2; MAGEC2; MAGE-C2; MAGE-C2 antigen; MAGEE1; MAGE-E1 antigen; melanoma antigen family C, 2; melanoma antigen family C2; melanoma antigen, family E, 1, cancer/testis specific; melanoma-associated antigen C2; uncharacterized protein LOC317590 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51438 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBF1 | |
| MAGEC2 | |
| A synthesized peptide derived from human MAGEC2(Accession Q9UBF1), corresponding to amino acid residues G194-A244. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction