missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGI2, Mouse anti-Human, Clone: 6C8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009863-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: PANSMVPPLAIMERPPPVMVNGRHNYETYLEYISRTSQSVPDITDRPPHSLHSMPTDGQLDGTYPPPVHDDNVSMASSGATQAELMTLTIVKGAQGFGFTIADSPTGQRVSpecifications
| MAGI2 | |
| Monoclonal | |
| Unconjugated | |
| membrane associated guanylate kinase, WW and PDZ domain containing 2 | |
| ACVRIP1/AIP1/ARIP1/MAGI-2/SSCAM | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 6C8 | |
| In 1x PBS, pH 7.4 | |
| NM_012301 | |
| MAGI2 | |
| MAGI2 (NP_036433, 519 a.a. ∼ 628 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9863 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction