missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564925
This item is not returnable.
View return policy
Description
Immunogen sequence: LKVGDHISAVN GQSIVELSHD NIVQLIKDAG VTVTLTVIAE EEHHGPPSGT NSARQSPALQ HRPMGQSQAN HIPGDRSALE GEIGKDVSTS YRHSWSDHKH LAQPDTAVIS VVGSRHNQNL GCYPVELERG PRGFGFSLRG Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.
Membrane-associated guanylate kinase with inverted orientation (MAGI3) is a novel, 130 kDa guanylate kinase which is part of a family of multi-PDZ domain containing guanylate kinases. These guanylate kinases are localized to epithelial cell tight junctions. MAGI3 and PTEN/MMAC cooperate to modulate the kinase activity of AKT/protein kinase B (PKB). MAGI3 positions the PTEN/MMAC phosphatase to specific subcellular locations that are involved with the regulation of cell proliferation and survival. MAGI3, as well as other proteins in this family, are made up of an N-terminal guanylate kinase domain, followed by a WW domain, named for two conserved tryptophan residues and first identified as two repeats in mouse YAP65, which is them followed by 5 PDZ domains.
Specifications
| MAGI3 | |
| Polyclonal | |
| Unconjugated | |
| MAGI3 | |
| 4732496O19Rik; 6530407C02Rik; AA407180; AI120132; dJ730K3.2; GKWW domain; KIAA1634; Magi3; MAGI-3; membrane associated guanylate kinase, WW and PDZ domain containing 3; membrane-associated guanylate kinase inverted 3; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 3; membrane-associated guanylate kinase-related (MAGI-3); membrane-associated guanylate kinase-related 3; mKIAA1634; PDZ 4-5; RP4-730K3.1; scaffolding protein SLIPR; scaffolding-like protein; Slipr | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 260425 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TCQ9 | |
| MAGI3 | |
| Recombinant protein fragment of Human MAGI3 (Product #RP-89519) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction