Learn More
Thermo Scientific™ Magnesium Oxide Rectangular Tray;Length (mm), 100;Width (mm), 25;Height (mm), 12.7
Brand:  Thermo Scientific™ 045634.KS
Product Code. 15474005
  • ONLINE EXCLUSIVE PRICE
    € 184.00
    € 180.00 / Each
    Save € 4.00
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 x 1 x 0.5 in. (100 x 25 x 12.7)
25 mm
Tray
1 Each
0.5 in.
1 in.
12.7 mm
Magnesium Oxide
100 mm
4 in.
Rectangular