Learn More
Brady™ Magnetic Plastic Transport Signs
Click to view available options
Displayed Text:
Flammable Solid 4
Flammable When Wet 4
Highly Flammable Liquid 3
Spontaneous Ignition 4
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | ISO 7010 |
| Material | Plastic |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Magnetic Plastic |
| Height (Metric) | 297 mm |
| Width (Metric) | 297 mm |
| Shape | Diamond |
| Displayed Text | Flammable When Wet 4 |
| Product Type | Transport Sign |
| Show More |