Learn More
Invitrogen™ MagniSort™ Mouse CD4 Positive Selection Kit
Product Code. 15521257
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Contains Sodium azide
Product Code. 15521257
Brand: Invitrogen™ 8802684174
Contains Sodium azide
Includes
  • MagniSort Anti-Mouse CD4 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A
Designed for the magnetic separation of CD4+ cells by positive selection. It has been optimized for the isolation of CD4+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD4 antibody and streptavidin-coated magnetic beads.
  • CD4+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD4+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD4, clones RM4-5 or GK1.5
TRUSTED_SUSTAINABILITY
Specifications
Cell Type CD4+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse