missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGOH Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97072AF350
This item is not returnable.
View return policy
Description
MAGOH Polyclonal specifically detects MAGOH in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MAGOH | |
| Polyclonal | |
| Western Blot, ELISA | |
| MAGOHAMAGOH1, mago-nashi (Drosophila) homolog, proliferation-associated, mago-nashi homolog, proliferation-associated (Drosophila), protein mago nashi homolog | |
| Produced in rabbits immunized with purified, recombinant MAGOH (NP_002361.1; Met1-Ile146). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 4116 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction