Learn More
Invitrogen™ Magrolimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558501
 View more versions of this product
Product Code. 30282917
  • € 477.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Magrolimab is an anti-CD47 monoclonal antibody designed to promote the phagocytosis of cancer cells by macrophages. Magrolimab is an anti-CD47 monoclonal antibody. Its purpose is to interfere with the recognition of CD47 by SIRPalpha receptors on macrophages, thereby blocking the self-protection signal used by cancer cells to avoid phagocytosis by macrophages.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Magrolimab Humanized
Recombinant Monoclonal
Unconjugated
Hu5F9-G4
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized