missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAK Antibody (OTI5A7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72580AF405
This item is not returnable.
View return policy
Description
MAK Monoclonal specifically detects MAK in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAK | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAK | |
| Recombinant protein fragment corresponding to amino acids 291-623 of human MAK (NP_005897) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5A7 | |
| Western Blot, Immunohistochemistry | |
| dJ417M14.2, EC 2.7.11, EC 2.7.11.22, male germ cell-associated kinaseserine/threonine protein kinase MAK, serine/threonine-protein kinase MAK | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 4117.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction