missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAK Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808770S
This item is not returnable.
View return policy
Description
MAK Monoclonal antibody specifically detects MAK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MAK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14893 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8487 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| GEMIN2 | |
| A930010O05Rik; dJ417M14.2; MAK; male germ cell associated kinase; male germ cell-associated kinase; male germ cell-associated kinase retinal-enriched isoform; protein kinase RCK; Rck; RP62; serine/threonine protein kinase MAK; serine/threonine-protein kinase MAK; testicular secretory protein Li 28; Unknown (protein for MGC:134523) | |
| Full length human recombinant protein of human GEMIN2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction