missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malaria pLDH Mouse anti-Protozoa, Clone: HM088, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13613.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against malaria pLDH.
- High specificity
- High reactivity
Specifications
| Malaria pLDH | |
| Monoclonal | |
| Unconjugated | |
| Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Lateral Flow | |
| HM088 | |
| Mouse monoclonal antibody raised against malaria pLDH. | |
| In 10 mM PBS, pH 7.4. | |
| Recombinant malaria pLDH antigen. | |
| RUO | |
| Protozoa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction