Learn More
Value Plastics Male Luer Slip Union Fittings
Shop All Cole-Parmer Products
Click to view available options
:
25 Luer Fitting Union White Nylon ML Slip
Description
Quick and easy assembly and disassembly without the need for tools. Constructed of durable chemical resistance nylon. Conforms to ISO-594-1 and ISO 594-2 standards
Spécification
Spécification
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Packaging Quantity | 25/Pk. |
| Inlet | Male Luer Slip |