Learn More
Malondialdehyde Antibody (11E3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59367FR
Product Code. 18225873
  • ONLINE EXCLUSIVE PRICE
    € 749.00
    € 707.70 / 100µL
    Save € 41.30
    5% Off
Description
Malondialdehyde Monoclonal specifically detects Malondialdehyde in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Malondialdehyde
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.
Store at 4C in the dark.
IgG1
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
11000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific