Learn More
Malondialdehyde Antibody (11E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59367
 View more versions of this product
Product Code. 18239273
  • € 690.00 / 100µg
Description
Malondialdehyde Monoclonal specifically detects Malondialdehyde in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Malondialdehyde
Monoclonal
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
Non-species specific
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
11000
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Mouse
Protein G purified
RUO
Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.
Store at -20C.