missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malondialdehyde Antibody (6H6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 194.00 - € 669.00
Specifications
| Antigen | Malondialdehyde |
|---|---|
| Clone | 6H6 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222955
|
Novus Biologicals
NBP2-59366 |
100 μg |
€ 669.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672767
|
Novus Biologicals
NBP2-59366-25ug |
25 μg |
€ 194.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Malondialdehyde Monoclonal specifically detects Malondialdehyde in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Malondialdehyde | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein G purified |
| 6H6 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title