Learn More
Invitrogen™ Malondialdehyde Monoclonal Antibody (6H6), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545804
Description
A 1:1,000 dilution of MA5-45804 was sufficient for detection of Malondialdehyde in 2 μg of Malondialdehyde conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.
Malondialdehyde is an organic compound with the formula CH2(CHO)2, and a byproduct of lipid metabolism in the body. Malondialdehyde, a highly reactive compound, is one of the many reactive electrophile species that cause toxic stress in cells and form covalent protein adducts, called advanced lipoxidation end products (ALE). This compound also forms mutagenic DNA adducts when it reacts with deoxyadenosine and deoxyguanosine in DNA. Malondialdehyde is also found in many foods and is present in high levels in rancid foods.
Specifications
| Malondialdehyde | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 6H6 | |
| PE | |
| 3-Propanedial; Malonaldehyde; Malondialdehyde; MDA | |
| Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.