missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MALSU1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562569

Product Code. 15760362

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GSAVGAEPGL RLLAVQRLPV GAAFCRACQT PNFVRGLHSE PGLEERAEGT VNEGRPESDA ADHTGPKFDI DMMVSLLRQE NARDICVIQV PPEMRYTDYF VIVS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.

Required for normal mitochondrial ribosome function and mitochondrial translation (PubMed:22238375, PubMed:23171548). May play a role in ribosome biogenesis by preventing premature association of the 28S and 39S ribosomal subunits (Probable). Interacts with mitochondrial ribosomal protein L14 (MRPL14), probably blocking formation of intersubunit bridge B8, preventing association of the 28S and 39S ribosomal subunits (Probable). Addition to isolated mitochondrial ribosomal subunits partially inhibits translation, probably by interfering with the association of the 28S and 39S ribosomal subunits and the formation of functional ribosomes (Probable). May also participate in the assembly and/or regulation of the stability of the large subunit of the mitochondrial ribosome (PubMed:22238376, PubMed:23171548). May function as a ribosomal silencing factor (Probable). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MALSU1
Polyclonal
Unconjugated
MALSU1
2410003K15Rik; C7orf30; Malsu1; Matsu1; mitochondrial assembly of ribosomal large subunit 1; mitochondrial assembly of ribosomal large subunit protein 1; mtRsfA; protein C7orf30 homolog; RGD1306936
Rabbit
Antigen Affinity Chromatography
RUO
115416
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96EH3
MALSU1
Recombinant protein corresponding to Human MALSU1. Recombinant protein control fragment (Product #RP-92768).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.