missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Antibody (MT1/3159R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08825JF646
This item is not returnable.
View return policy
Description
MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MALT1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase | |
| Human MALT1 recombinant fragment (aa701-808) (Uniprot: Q9UDY8) | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10892 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| MT1/3159R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction