missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Monoclonal Antibody (OTI6D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807322S
This item is not returnable.
View return policy
Description
MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Western BlotSpecifications
| MALT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UDY8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10892 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6D2 | |
| Unconjugated | |
| MALT1 | |
| A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1 | |
| Full length human recombinant protein of human MALT1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction