Learn More
MALT1 Mouse anti-Human, Clone: OTI3E10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807328
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma. MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.Specifications
| MALT1 | |
| Monoclonal | |
| Unconjugated | |
| MALT1 | |
| IMD12, MLT, MLT1, PCASP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10892 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| OTI3E10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UDY8 | |
| MALT1 | |
| Full length human recombit protein of human MALT1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.