missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11660-1-AP-150UL
This item is not returnable.
View return policy
Description
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma. MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.Specifications
| MALT1 | |
| Polyclonal | |
| Unconjugated | |
| MALT1 | |
| MALT1, MLT, MLT1, Paracaspase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10892, 240354, 307366 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q2TBA3, Q9UDY8 | |
| Malt1 | |
| MALT1 Fusion Protein Ag2182 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction