missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Antibody (R29), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66609AF750
This item is not returnable.
View return policy
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Bacteria samples. It is validated for Western Blot.
Specifications
| Maltose Binding Protein | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
| Recombinant maltose binding protein | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| R29 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction