missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAMDC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566300
This item is not returnable.
View return policy
Description
Immunogen sequence: VCNFVCDCRDC SDEAQCGYHG ASPTLGAPFA CDFEQDPCGW RDISTSGYSW LRDRAGAALE GPGPHSDHTL GTDL Highest antigen sequence identity to the following orthologs - mouse 77%, rat 79%.
Probably involved in the sorting and selective transport of receptors and ligands across polarized epithelia. Target protein is a glycoprotein localized to the apical endosomal tubules of intestinal epithelial cells; may play a role in intestinal transport.
Specifications
| MAMDC4 | |
| Polyclonal | |
| Unconjugated | |
| MAMDC4 | |
| AEGP; apical early endosomal glycoprotein; apical endosomal glycoprotein; endotubin; MAM domain containing 4; MAM domain-containing protein 4; MAMDC4; UNQ3001/PRO9742 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 158056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXC1 | |
| MAMDC4 | |
| Recombinant Human MAMDC4. Recombinant protein control fragment (Product #RP-106980). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction