missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAML2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116738
This item is not returnable.
View return policy
Description
Positive Samples: A-549, 293T, Mouse brain, Mouse lung Immunogen sequence: NQQLMGKKQT LQRQIMEQKQ QLLLQQQMLA DAEKIAPQDQ INRHLSRPPP DYKDQRRNVG NMQPTAQYSG GSSTISLNSN QALANPVSTH TILTPNSSLL S.
Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
Specifications
| MAML2 | |
| Polyclonal | |
| Unconjugated | |
| MAML2 | |
| 5930431H10; BC032967; KIAA1819; MAM2; Mam-2; MAM3; MAM-3; MAML2; mastermind like 2 (Drosophila); mastermind like transcriptional coactivator 2; mastermind-like 2; mastermind-like protein 2; mastermind-like transcriptional coactivator 2; MLL-MAML2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 270118, 689844, 84441 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.18 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8IZL2 | |
| MAML2 | |
| A synthetic peptide corresponding to a sequence within amino acids 750-850 of human MAML2 (NP_1158031). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction