missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML3 Antibody (MAML3/1303), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54309H
This item is not returnable.
View return policy
Description
MAML3 Monoclonal antibody specifically detects MAML3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MAML3 | |
| Monoclonal | |
| HRP | |
| CAG/CTG 3, ERDA3, MAM-2, mam-3, mastermind-like 3 (Drosophila), mastermind-like protein 3, polyglutamine rich, trinucleotide repeat containing 3 | |
| Recombinant human MAML3 protein fragment (Uniprot: Q96JK9) | |
| 100 μg | |
| Neuroscience | |
| 55534 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MAML3/1303 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction