missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML3 Mouse, Clone: MAML3/1303, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 510.00
Specifications
Antigen | MAML3 |
---|---|
Clone | MAML3/1303 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940718
|
EnquireBio
55534-MSM1-P1 |
100 μg |
€ 510.00
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930718
|
EnquireBio
55534-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MAML3 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
55534 | |
Recombinant human full-length MAML3 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
150-170kDa |
MAML3/1303 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_061187 | |
MAML3 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
MCF-7 cells. Pancreas or Placenta. Cervix or Colon Carcinoma. |