missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (EPR9092), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-41752-500UL
This item is not returnable.
View return policy
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Mammaglobin A | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant | |
| SCGB2A2 | |
| A recombinant protein fragment corresponding to human Mammaglobin A. | |
| 4250 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| EPR9092 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Rabbit | |
| 500 μL | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction