missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (MGB/2704), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08429JF646
This item is not returnable.
View return policy
Description
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Mammaglobin A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Recombinant full-length human Mammaglobin A protein (Uniprot: Q13296) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| MGB/2704 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4250 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction