missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mammaglobin A Recombinant Mouse Monoclonal Antibody (A9A2-R)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550661
This item is not returnable.
View return policy
Description
Positive control: Human skin tissue, human breast tissue. Predicted band size: 10 kDa Subcellular Location: Secreted.
Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.
Specifications
| Mammaglobin A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q13296 | |
| SCGB2A2 | |
| Synthetic peptide within Human Mammaglobin A aa 21-70/93. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| A9A2-R | |
| Unconjugated | |
| SCGB2A2 | |
| C3.1; HGNC:7050; mammaglobin 1; mammaglobin A; Mammaglobin-1; Mammaglobin-A; MGB1; MGC71974; prostatein; prostatein C3 subunit; prostatein peptide C3; prostatic steroid binding protein, allele C3(1); prostatic steroid-binding protein C3; Psbpc3; SCGB2A2; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2; UGB2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4250 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction