missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAMSTR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69204
This item is not returnable.
View return policy
Description
MAMSTR Polyclonal specifically detects MAMSTR in Human samples. It is validated for Western Blot.
Specifications
| MAMSTR | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MAMSTR | |
| Synthetic peptides corresponding to FLJ36070 (MEF2 activating motif and SAP domain containing transcriptional regulator) The peptide sequence was selected from the middle region of FLJ36070. Peptide sequence PRAADTPGTAPAPTPTPAPAAAPALTPSSGPGSAALTLEEELQEAIRRAQ The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 284358 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| FLJ36070, MASTRlikely ortholog of MEF2-activating SAP transcriptional regulator, MEF2 activating motif and SAP domain containing transcriptional regulator, MEF2-activating motif and SAP domain-containing transcriptional regulator, MEF2-activating SAP transcriptional regulator, MEF2-activating SAP transcriptional regulatory protein, MGC117259 | |
| Rabbit | |
| 33 kDa | |
| 100 μL | |
| Primary | |
| This product recognizes Human. Zebrafish: 85%; Goat: 83%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Goat, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction