missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1C1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00057134-B01P
This item is not returnable.
View return policy
Description
MAN1C1 Polyclonal antibody specifically detects MAN1C1 in Human samples. It is validated for Western Blot
Specifications
| MAN1C1 | |
| Polyclonal | |
| Western Blot | |
| NP_065112.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| 1,2-alpha-mannosidase IC, Alpha-1,2-mannosidase IC, EC 3.2.1.113, HMICProcessing alpha-1,2-mannosidase IC, MAN1A3, MAN1C, Mannosidase alpha class 1C member 1, mannosidase, alpha, class 1C, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IC, pp6318 | |
| MAN1C1 (NP_065112.1, 1 a.a. - 630 a.a.) full-length human protein. MLMRKVPGFVPASPWGLRLPQKFLFLLFLSGLVTLCFGALFLLPHSSRLKRLFLAPRTQQPGLEVVAEIAGHAPAREQEPPPNPAPAAPAPGEDDPSSWASPRRRKGGLRRTRPTGPREEATAARGNSIPASRPGDEGVPFRFDFNAFRSRLRHPVLGTRADESQEPQSQVRAQREKIKEMMQFAWQSYKRYAMGKNELRPLTKDGYEGNMFGGLSGATVIDSLDTLYLMELKEEFQEAKAWVGESFHLNVSGEASLFEVNIRYIGGLLSAFYLTGEEVFRIKAIRLGEKLLPAFNTPTGIPKGVVSFKSGNWGWATAGSSSILAEFGSLHLEFLHLTELSGNQVFAEKVRNIRKVLRKIEKPFGLYPNFLSPVSGNWVQHHVSVGGLGDSFYEYLIKSWLMSGKTDMEAKNMYYEALEAIETYLLNVSPGGLTYIAEWRGGILDHKMGHLACFSGGMIALGAEDAKEEKRAHYRELAAQITKTCHESYARSDTKLGPEAFWFNSGREAVATQLSESYYILRPEVVESYMYLWRQTHNPIYREWGWEVVLALEKYCRTEAGFSGIQDVYSSTPNHDNKQQSFFLAETLKYLYLLFSEDDLLSLEDWVFNTEAHPLPVNHSDSSGRAWGRH | |
| 0.05 mg | |
| Stem Cell Markers | |
| 57134 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction