missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN2A2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98095JF646
This item is not returnable.
View return policy
Description
MAN2A2 Polyclonal antibody specifically detects MAN2A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MAN2A2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| alpha-mannosidase 2x, alpha-mannosidase IIx, MAN IIx, MANA2X, mannosidase, alpha type II-X, mannosidase, alpha, class 2A, member 2, mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase, manosidase, alpha-, type II, isozyme X | |
| Produced in rabbits immunized with E. coli-derived Human MAN2A2 fragment. | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers | |
| 4122 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction