missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAN2B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557911
This item is not returnable.
View return policy
Description
Immunogen sequence: LLCHRIEQEY QAGPLELNRE AVLRTSTNLN SQQVIYSDNN GYQMQRRPYV SYVNNSIARN YYPMVQSAFM EDGKSRLVLL SERAHGISSQ GNGQVE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
The MAN2B2 (Mannosidase Alpha Class 2B Member 2) gene encodes an enzyme that is involved in the processing and modification of glycoproteins by hydrolyzing alpha-mannosidic linkages. Located on chromosome 4, the MAN2B2 protein is crucial for proper lysosomal function and is part of the glycosyl hydrolase family. Pathogenic variants in the MAN2B2 gene have been linked to a rare congenital disorder of glycosylation (CDG), known as MAN2B2-CDG, characterized by immune deficiency, developmental delay, stroke, and dysmorphic facial features. These mutations lead to an altered mannosylation profile, impacting glycoprotein processing and cellular functions. Studies have shown that reintroducing wild-type MAN2B2 can restore the normal mannosylation profile in patient cells, highlighting its therapeutic potential. Furthermore, recent research has identified circular RNA from this gene (circMAN2B2) that is implicated in the proliferation of hepatocellular carcinoma cells, suggesting a role in cancer biology.
Specifications
| MAN2B2 | |
| Polyclonal | |
| Unconjugated | |
| MAN2B2 | |
| 135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Ab2-450; core-specific lysosomal alpha-1,6-Mannosidase; epididymis-specific alpha-mannosidase; HMG17L3; Kiaa0935; Man2b2; mannosidase 2, alpha B2; mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; mKIAA0935; NHC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2E5 | |
| MAN2B2 | |
| Recombinant protein corresponding to Human MAN2B2. Recombinant protein control fragment (Product #RP-96590). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction