missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAN2B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113537
This item is not returnable.
View return policy
Description
MAN2B2 Polyclonal Antibody for ELISA, IHC (P), Western Blot
The MAN2B2 (Mannosidase Alpha Class 2B Member 2) gene encodes an enzyme that is involved in the processing and modification of glycoproteins by hydrolyzing alpha-mannosidic linkages. Located on chromosome 4, the MAN2B2 protein is crucial for proper lysosomal function and is part of the glycosyl hydrolase family. Pathogenic variants in the MAN2B2 gene have been linked to a rare congenital disorder of glycosylation (CDG), known as MAN2B2-CDG, characterized by immune deficiency, developmental delay, stroke, and dysmorphic facial features. These mutations lead to an altered mannosylation profile, impacting glycoprotein processing and cellular functions. Studies have shown that reintroducing wild-type MAN2B2 can restore the normal mannosylation profile in patient cells, highlighting its therapeutic potential. Furthermore, recent research has identified circular RNA from this gene (circMAN2B2) that is implicated in the proliferation of hepatocellular carcinoma cells, suggesting a role in cancer biology.
Specifications
| MAN2B2 | |
| Polyclonal | |
| Unconjugated | |
| MAN2B2 | |
| 135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Ab2-450; core-specific lysosomal alpha-1,6-Mannosidase; epididymis-specific alpha-mannosidase; HMG17L3; Kiaa0935; Man2b2; mannosidase 2, alpha B2; mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; mKIAA0935; NHC | |
| Rabbit | |
| Protein G | |
| RUO | |
| 23324 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9Y2E5 | |
| MAN2B2 | |
| Recombinant Human Epididymis-specific alpha-mannosidase protein (347-523AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction