missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANF Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-06020
This item is not returnable.
View return policy
Description
MANF Polyclonal specifically detects MANF in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| ARMET | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| arginine-rich, mutated in early stage tumors, ARMETArginine-rich protein, ARPMGC142150, mesencephalic astrocyte-derived neurotrophic factor, MGC142148, Protein ARMET | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_006001.2 | |
| MANF | |
| Peptide with sequence KFCREARGKENR corresponding to internal region according to NP_006001.2. | |
| 0.1 mg | |
| Cancer, DNA Repair | |
| 7873 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction