missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC5L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85719
This item is not returnable.
View return policy
Description
CDC5L Polyclonal specifically detects CDC5L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CDC5L | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CDC5, CDC5 (cell division cycle 5, S. pombe, homolog)-like, CDC5 cell division cycle 5-like (S. pombe), Cdc5-like protein, Cdc5-related protein, CEF1, cell division cycle 5-like protein, dJ319D22.1, dJ319D22.1 (CDC5-like protein), hCDC5, KIAA0432, PCDC5RPCDC5-LIKE, Pombe cdc5-related protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 988 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CDC5L | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DLQKSEELIKKEMITMLHYDLLHHPYEPSGNKKGKTVGFGTNNSEHITYLEHNPYEKFSKEELKKAQDVLVQEMEVVKQGMSHGELSSEAYNQVWEECYSQVLYLPGQSRYTRANLASKKDRIESLEKRLEINRGHM | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction