missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Manganese Reagent Powder Pillow Set
Click to view available options
Range:
0.1 to 20.0mg/L Mn
Unit Size:
Each
Description
Powder Pillows make titration applications quick and easy.
- Sealed packets ensure fresh, uncontaminated reagent every time
- Premeasured to avoid measurement error and contamination from handling
- Ready to use packets save time and hassle
- Exact doses to easily monitor supply and prepare for lab procedures
- Single-use packs for easy transportation and packing in test kits
Specifications
Specifications
| Analytes | Manganese |
| Certifications/Compliance | US EPA |
| Format | Individual Packet |
| For Use With (Application) | Manganese determination |
| For Use With (Equipment) | All except DR/3000, DR/2000 |
| Includes | Citrate Buffer Powder Pillows and Sodium Periodate Powder Pillows |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 0.1 to 20.0mg/L Mn |
| Sample Size | 10 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction