missing translation for 'onlineSavingsMsg'
Learn More

Manganese Standard: Mn @ 10000 μg/mL in 5% HNO3

Product Code. 30317311
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317311 250 mL -
30319640 100 mL 100mL
30325957 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30317311 Supplier VHG Labs Supplier No. VHGTMNN250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Manganese
CAS 7439-96-5
Analyte or Component Names Manganese
Shipping Condition Room Temperature
Molecular Formula Mn
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Mn
InChI Formula InChI=1 S/Mn
Solution Type Single Solution
SMILES [Mn]
Element Mn
IUPAC Name Manganese
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 54.94
Quantity 250 mL
Formula Weight 54.938
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.